Gene Transcripts

Gene Symbol
Fmo3
Entrez Gene ID
14262
Gene Name
flavin containing monooxygenase 3
Gene Aliases
AW111792
Chromosome Location
Chr. 1: 162953800 - 162984491 on Build GRCm38
UniGene ID
Mm.2900
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008030.1 NP_032056.1 9 1381
GenBank mRNA AK149440.1 - 9 1408
U87147.1 - 9 1381

View Full Product Details