Gene Transcripts

Gene Symbol
H2-Ob
Entrez Gene ID
15002
Gene Name
histocompatibility 2, O region beta locus
Gene Aliases
A-beta-2, A-beta2, H-2Ab, H-2I, H-2Ob, H2-Ab, H2-Ab2, H2-IAb2, Ob
Chromosome Location
Chr. 17: 34238895 - 34245908 on Build GRCm38
UniGene ID
Mm.327358
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010389.3 NP_034519.2 4 722
GenBank mRNA AK053233.1 - 4 771
KJ650230.1 - 4 655
KJ663722.1 - 4 577

View Full Product Details