Gene Transcripts

Gene Symbol
Itga2b
Entrez Gene ID
16399
Gene Name
integrin alpha 2b
Gene Aliases
AI172977, CD41, CD41B, GpIIb, alphaIIb
Chromosome Location
Chr. 11: 102453297 - 102469883 on Build GRCm38
UniGene ID
Mm.26646
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010575.2 NP_034705.2 22, 23 2442
GenBank mRNA AF170316.1 - 22, 23 2303
AK048700.1 - 17, 18 1950
AK140749.1 - 17, 18 2094
AK142289.1 - 22, 23 2304
AK154619.1 - 22, 23 2443
AK156872.1 - 21, 22 2623
AK157012.1 - 21, 22 2627
AK158443.1 - 22, 23 2294
AK161727.1 - 2, 3 106
BC120493.1 - 22, 23 2292 40129416
BC137570.1 - 22, 23 2292 8860589

View Full Product Details