Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001277970.1 | NP_001264899.1 | 9 | 1489 | |
NM_008397.4 | NP_032423.2 | 9 | 1489 | ||
GenBank mRNA | AK034186.1 | - | 9 | 1346 | |
AK045391.1 | - | 2 | 70 | ||
AK159653.1 | - | 4 | 425 | ||
BC049132.1 | - | 10 | 1414 | ||
BC058095.1 | - | 10 | 1414 | 6826609 | |
X69902.1 | - | 9 | 1470 |