Gene Transcripts

Gene Symbol
Slc2a10
Entrez Gene ID
170441
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 10
Gene Aliases
AA450473, Glut10
Chromosome Location
Chr. 2: 165503897 - 165519917 on Build GRCm38
UniGene ID
Mm.34147
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130451.3 NP_569718.1 2 1278
GenBank mRNA AK052156.1 - 2 1245
AY029579.1 - 2 1201
BC137917.1 - 2 1196 8860936
BC137920.1 - 2 1196 8860939

View Full Product Details