Gene Transcripts

Gene Symbol
Pah
Entrez Gene ID
18478
Gene Name
phenylalanine hydroxylase
Gene Aliases
AW106920
Chromosome Location
Chr. 10: 87521795 - 87584137 on Build GRCm38
UniGene ID
Mm.263539
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008777.3 NP_032803.2 5 700
GenBank mRNA AK145839.1 - 5 565
AK149519.1 - 5 566
BC013458.1 - 5 558 4223155
X51942.1 - 5 525

View Full Product Details