Gene Transcripts

Gene Symbol
Prkcg
Entrez Gene ID
18752
Gene Name
protein kinase C, gamma
Gene Aliases
PKCgamma, Pkcc, Prkcc
Chromosome Location
Chr. 7: 3303532 - 3331099 on Build GRCm38
UniGene ID
Mm.7980
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291434.1 NP_001278363.1 13 1564
NM_011102.4 NP_035232.1 14 1717
GenBank mRNA AK144419.1 - 14 1719
BC111807.1 - 13 1451 40039740
L28035.1 - 14 1795
X67129.1 - 14 1526

View Full Product Details