Gene Transcripts

Gene Symbol
Rph3a
Entrez Gene ID
19894
Gene Name
rabphilin 3A
Gene Aliases
2900002P20Rik, AU022689, AW108370
Chromosome Location
Chr. 5: 120940499 - 121010102 on Build GRCm38
UniGene ID
Mm.181166
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB112924.1 - 5 450
AK139040.1 - 4 392
BC042585.1 - 5 617 5364086
BC050883.1 - 5 584 5698459
D29965.1 - 5 427

View Full Product Details