Gene Transcripts

Gene Symbol
Tyrp1
Entrez Gene ID
22178
Gene Name
tyrosinase-related protein 1
Gene Aliases
Oca3, TRP-1, TRP1, Tyrp, b, brown, isa
Chromosome Location
Chr. 4: 80834208 - 80851736 on Build GRCm38
UniGene ID
Mm.30438
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282014.1 NP_001268943.1 7 1652
NM_001282015.1 NP_001268944.1 7 1646
NM_031202.3 NP_112479.1 7 1667
GenBank mRNA AK148332.1 - 7 1652
AK148370.1 - 7 1558
AK148441.1 - 7 1558
AK177043.1 - 2 153
AK178803.1 - 2 153
AK181989.1 - 2 151
AK188091.1 - 2 153
AK189488.1 - 2 153
AK190379.1 - 1 133
AK192440.1 - 2 153
AK193585.1 - 2 153
AK194794.1 - 1 130
AK194981.1 - 1 133
AK196132.1 - 1 130
AK197740.1 - 2 153
AK198402.1 - 1 143
AK198835.1 - 2 153
AK198900.1 - 1 135
AK199149.1 - 2 153
AK199163.1 - 2 153
AK199776.1 - 2 153
AK199872.1 - 2 153
AK200013.1 - 2 153
AK200321.1 - 2 137
AK200365.1 - 2 153
AK200395.1 - 2 153
AK201323.1 - 1 119
AK203101.1 - 2 153
AK203562.1 - 1 108
AK204301.1 - 2 145
AK207921.1 - 2 166
BC076598.1 - 7 1650 30613975
X03687.1 - 7 1552

View Full Product Details