Gene Transcripts

Gene Symbol
Uba1
Entrez Gene ID
22201
Gene Name
ubiquitin-like modifier activating enzyme 1
Gene Aliases
A1S9, Sbx, Ube-1, Ube1x
Chromosome Location
Chr. X: 20658302 - 20683179 on Build GRCm38
UniGene ID
Mm.1104
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136085.2 NP_001129557.1 20 2562
NM_001276316.1 NP_001263245.1 21 2654
NM_001276317.1 NP_001263246.1 20 2557
NM_009457.4 NP_033483.2 20 2557
GenBank mRNA AK081808.1 - 20 2531
AK088057.1 - 20 2439
AK088528.1 - 20 2439
AK143416.1 - 20 2463
AK171667.1 - 20 2528
BC058630.1 - 20 2503 6402368
BC138200.1 - 20 2420 8861219
BC145984.1 - 20 2421 40131287
D10576.1 - 20 2423
X62580.1 - 5 492

View Full Product Details