Gene Transcripts

Gene Symbol
Smg5
Entrez Gene ID
229512
Gene Name
Smg-5 homolog, nonsense mediated mRNA decay factor (C. elegans)
Gene Aliases
BC024683, mKIAA1089
Chromosome Location
Chr. 3: 88336260 - 88362338 on Build GRCm38
UniGene ID
Mm.439830
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178246.3 NP_839977.2 10 1256
GenBank mRNA AK129286.1 - 10 1235
AK133688.1 - 1 80
AK143911.1 - 10 1258
AK155787.1 - 10 1245
AK172217.1 - 9 1136
BC024683.1 - 10 1208 3992000
BC066787.1 - 4 483
BC100408.1 - 10 1205 30734268

View Full Product Details