Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172701.2 | NP_766289.2 | 4, 5 | 630 | |
GenBank mRNA | AB154826.1 | - | 4, 5 | 583 | |
AB286960.1 | - | 5, 6 | 631 | ||
AK077467.1 | - | 4, 5 | 596 | ||
AK077909.1 | - | 4, 5 | 551 | ||
BC045150.1 | - | 4, 5 | 576 | 6309923 |