Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145589.2 | NP_663564.2 | 2 | 405 | |
GenBank mRNA | AK133884.1 | - | 2 | 370 | |
AK139559.1 | - | 3 | 180 | ||
AK145816.1 | - | 3 | 185 | ||
AK151203.1 | - | 3 | 237 | ||
BC055033.1 | - | 2 | 405 | 6310272 |