Gene Transcripts

Gene Symbol
Gpatch8
Entrez Gene ID
237943
Gene Name
G patch domain containing 8
Gene Aliases
5430405G24Rik, AU018890, ENSMUSG00000075516, Fbm1, Gpatc8, mKIAA0553
Chromosome Location
Chr. 11: 102475911 - 102556158 on Build GRCm38
UniGene ID
Mm.260282
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159492.1 NP_001152964.1 8 3251
GenBank mRNA AK122307.1 - 1 701
AK142570.1 - 1 495
BC035255.1 - 6 3310

View Full Product Details