Gene Transcripts

Gene Symbol
Arid4a
Entrez Gene ID
238247
Gene Name
AT rich interactive domain 4A (RBP1-like)
Gene Aliases
A630009N03, A630067N03Rik, MmRBBP1, Rbbp1
Chromosome Location
Chr. 12: 71015967 - 71099351 on Build GRCm38
UniGene ID
Mm.241601
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081195.1 NP_001074664.1 15 1448
GenBank mRNA AK050594.1 - 14 1255
AK149171.1 - 5 508
AY116674.1 - 15 1334
BC025436.1 - 15 1436
BC058209.1 - 15 1365
BC080780.1 - 15 1383
BC157975.1 - 14 1250 9007297
BC158109.1 - 14 1250 9007390

View Full Product Details