Gene Transcripts

Gene Symbol
Parp12
Entrez Gene ID
243771
Gene Name
poly (ADP-ribose) polymerase family, member 12
Gene Aliases
9930021O16, AA409132, AA536654, ARTD12, PARP-12, Zc3hdc1
Chromosome Location
Chr. 6: 39086412 - 39118349 on Build GRCm38
UniGene ID
Mm.268462
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172893.3 NP_766481.2 6 1360
GenBank mRNA AK036886.1 - 6 1363
AK053205.1 - 6 1261
AK081528.1 - 6 1261
AK156623.1 - 6 1229
AK162200.1 - 6 931
AK204869.1 - 3 297
BC048927.1 - 2 211
BC120733.1 - 6 1193 40129656
BC137645.1 - 6 1193 8860664

View Full Product Details