Gene Transcripts

Gene Symbol
Parp12
Entrez Gene ID
243771
Gene Name
poly (ADP-ribose) polymerase family, member 12
Gene Aliases
9930021O16, AA409132, AA536654, ARTD12, PARP-12, Zc3hdc1
Chromosome Location
Chr. 6: 39086412 - 39118349 on Build GRCm38
UniGene ID
Mm.268462
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172893.3 NP_766481.2 12 2059
GenBank mRNA AK036886.1 - 12 2062
AK053205.1 - 12 1960
AK081528.1 - 12 1964
AK156623.1 - 12 1928
AK162200.1 - 12 1631
BC024579.1 - 7 708
BC028906.1 - 4 418
BC048927.1 - 8 910
BC120733.1 - 12 1892 40129656
BC137645.1 - 12 1892 8860664

View Full Product Details