Gene Transcripts

Gene Symbol
Arhgap20
Entrez Gene ID
244867
Gene Name
Rho GTPase activating protein 20
Gene Aliases
6530403F17Rik, A530023E23Rik, RARhoGAP, mKIAA1391
Chromosome Location
Chr. 9: 51765087 - 51853856 on Build GRCm38
UniGene ID
Mm.400352
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175535.3 NP_780744.2 11 1399
GenBank mRNA AK129345.1 - 8 888

Gene Symbol
Arhgap20os
Entrez Gene ID
408028
Gene Name
Rho GTPase activating protein 20, opposite strand
Gene Aliases
9230115E21Rik
Chromosome Location
Chr. 9: 51839493 - 51875885 on Build GRCm38
UniGene ID
Mm.393649
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK033819.1 - 4 399
AK135715.1 - 454
AK165145.1 - 3 418

View Full Product Details