Gene Transcripts

Gene Symbol
Clspn
Entrez Gene ID
269582
Gene Name
claspin
Gene Aliases
B130025E01, C85083, E130314M08Rik
Chromosome Location
Chr. 4: 126556951 - 126594533 on Build GRCm38
UniGene ID
Mm.243962
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175554.4 NP_780763.3 14 2566
GenBank mRNA AK136174.1 - 7 1414
BC050848.1 - 14 2521 6507692

View Full Product Details