Gene Transcripts

Gene Symbol
Abcc6
Entrez Gene ID
27421
Gene Name
ATP-binding cassette, sub-family C (CFTR/MRP), member 6
Gene Aliases
Abcc1b, DCC, Dyscalc1, Mrp6, dyscalc
Chromosome Location
Chr. 7: 45974115 - 46030486 on Build GRCm38
UniGene ID
Mm.63514
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018795.2 NP_061265.2 30, 31 4391
GenBank mRNA AB028737.1 - 30, 31 4397
BC040400.1 - 30, 31 4399
BC049980.1 - 14, 15 2334

View Full Product Details