Gene Transcripts

Gene Symbol
Slc15a5
Entrez Gene ID
277898
Gene Name
solute carrier family 15, member 5
Gene Aliases
9830102E05Rik
Chromosome Location
Chr. 6: 137983590 - 138079916 on Build GRCm38
UniGene ID
Mm.269823
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177787.4 NP_808455.2 4 817
GenBank mRNA AK036413.1 - 3 600
BC141332.1 - 3 536 9055944
BC141334.1 - 3 536 9055946

View Full Product Details