Gene Transcripts

Gene Symbol
Bcar3
Entrez Gene ID
29815
Gene Name
breast cancer anti-estrogen resistance 3
Gene Aliases
AI131758, AND-34
Chromosome Location
Chr. 3: 122419780 - 122530189 on Build GRCm38
UniGene ID
Mm.45815
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013867.2 NP_038895.1 10, 11 2483
GenBank mRNA AF179566.1 - 10, 11 2431
AK143894.1 - 10, 11 2271
AK158241.1 - 10, 11 2484
AK167578.1 - 10, 11 2301
BC023930.1 - 10, 11 2472 5324365

View Full Product Details