Gene Transcripts

Gene Symbol
Pitpnm3
Entrez Gene ID
327958
Gene Name
PITPNM family member 3
Gene Aliases
A330068P14Rik, AI848332, Ackr6, Gm880
Chromosome Location
Chr. 11: 72047528 - 72135889 on Build GRCm38
UniGene ID
Mm.351793
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024927.2 NP_001020098.1 12 1760
NM_001081641.1 NP_001075110.1 11 1712
GenBank mRNA AK147444.1 - 12 1717
AK147630.1 - 11 1545

View Full Product Details