Gene Transcripts

Gene Symbol
Cdkl4
Entrez Gene ID
381113
Gene Name
cyclin-dependent kinase-like 4
Gene Aliases
AU067824, Gm942
Chromosome Location
Chr. 17: 80523550 - 80577839 on Build GRCm38
UniGene ID
Mm.329216
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033443.4 NP_001028615.1 3 418

View Full Product Details