Gene Transcripts

Gene Symbol
Slc4a5
Entrez Gene ID
232156
Gene Name
solute carrier family 4, sodium bicarbonate cotransporter, member 5
Gene Aliases
C330006M16, C330016K18Rik
Chromosome Location
Chr. 6: 83207580 - 83304945 on Build GRCm38
UniGene ID
Mm.333189
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166067.1 NP_001159539.1 9 1273
GenBank mRNA CO431509.1 - 4 323
CX562762.1 - 6 585

View Full Product Details