Gene Transcripts
- Gene Symbol
- Ush2a
- Entrez Gene ID
- 22283
- Gene Name
- Usher syndrome 2A (autosomal recessive, mild)
- Gene Aliases
- A930011D15Rik, A930037M10Rik, Gm676, Gm983, Mush2a, Usherin
- Chromosome Location
- Chr. 1: 188262017 - 188965497 on Build GRCm38
- UniGene ID
- Mm.331238
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021408.3 |
NP_067383.3
|
38, 39 |
7609 |
|
GenBank mRNA |
DQ073638.1 |
- |
38, 39 |
7413 |
|
View Full Product Details