Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009455.3 | NP_033481.1 | 4, 5 | 528 | |
GenBank mRNA | AK090177.1 | - | 4, 5 | 529 | |
AK148266.1 | - | 4, 5 | 459 | ||
BC003781.1 | - | 4, 5 | 445 | 3484294 | |
X92665.1 | - | 4, 5 | 490 |