Gene Transcripts
- Gene Symbol
- Arhgef5
- Entrez Gene ID
- 54324
- Gene Name
- Rho guanine nucleotide exchange factor (GEF) 5
- Gene Aliases
- 2210412D05Rik, AA717842, AW495314, Tim1
- Chromosome Location
- Chr. 6: 43265644 - 43289320 on Build GRCm38
- UniGene ID
- Mm.320978
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_133674.1 |
NP_598435.1
|
2 |
2609 |
|
View Full Product Details