Gene Transcripts

Gene Symbol
Dfna5
Entrez Gene ID
54722
Gene Name
deafness, autosomal dominant 5 (human)
Gene Aliases
2310037D07Rik, 4932441K13Rik, Dfna5h, EG14210, Fin15
Chromosome Location
Chr. 6: 50207403 - 50262862 on Build GRCm38
UniGene ID
Mm.248361
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018769.3 NP_061239.1 3, 4 463
GenBank mRNA AF073309.1 - 3, 4 464
AK016561.1 - 831
AK089459.1 - 450
AK155135.1 - 442
AK171278.1 - 3, 4 462
BC051962.1 - 3, 4 523
BC086618.1 - 3, 4 523
BC132303.1 - 2, 3 434 40130580

View Full Product Details