Gene Transcripts

Gene Symbol
Pdss1
Entrez Gene ID
56075
Gene Name
prenyl (solanesyl) diphosphate synthase, subunit 1
Gene Aliases
2610203G20Rik, 2700031G06Rik, TPT, TPT 1, Tprt, mDLP1, mSPS1
Chromosome Location
Chr. 2: 22893560 - 22940266 on Build GRCm38
UniGene ID
Mm.249752
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019501.3 NP_062374.2 5 624
GenBank mRNA AB210841.1 - 5 544
AF118855.1 - 3 319
AK012307.1 - 5 552
BC026820.1 - 5 526 4039464
BC107273.1 - 5 439
BC107274.1 - 5 443

View Full Product Details