Gene Transcripts
- Gene Symbol
- Snx1
- Entrez Gene ID
- 56440
- Gene Name
- sorting nexin 1
- Gene Aliases
- Chromosome Location
- Chr. 9: 66088127 - 66124886 on Build GRCm38
- UniGene ID
- Mm.271891
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019727.2 |
NP_062701.2
|
7, 8 |
846 |
|
GenBank mRNA |
AB019214.1 |
- |
7, 8 |
786 |
|
AF154120.1 |
- |
7, 8 |
850 |
|
AK149850.1 |
- |
7, 8 |
758 |
|
AK154079.1 |
- |
7, 8 |
759 |
|
AK155746.1 |
- |
7, 8 |
734 |
|
AK161241.1 |
- |
7, 8 |
827 |
|
AK167177.1 |
- |
7, 8 |
801 |
|
AK167993.1 |
- |
7, 8 |
741 |
|
AK168742.1 |
- |
7, 8 |
839 |
|
AK177911.1 |
- |
1, 2 |
36 |
|
AK189960.1 |
- |
1, 2 |
36 |
|
AK192993.1 |
- |
1, 2 |
36 |
|
BC066044.1 |
- |
7, 8 |
803 |
6807363 |
BC066189.1 |
- |
7, 8 |
737 |
30471747 |
View Full Product Details