Gene Transcripts
- Gene Symbol
- Ttyh1
- Entrez Gene ID
- 57776
- Gene Name
- tweety homolog 1 (Drosophila)
- Gene Aliases
- 4930459B04Rik, 6330408P11Rik, TTY1, mTTY1, tty
- Chromosome Location
- Chr. 7: 4119527 - 4136241 on Build GRCm38
- UniGene ID
- Mm.29729
- Species
- Mus musculus
- Gene Symbol
- D030047H15Rik
- Entrez Gene ID
- 100037396
- Gene Name
- RIKEN cDNA D030047H15 gene
- Gene Aliases
- Gm15924, OTTMUSG00000027889
- Chromosome Location
- Chr. 7: 4126340 - 4136926 on Build GRCm38
- UniGene ID
- Mm.447887
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK050966.1 |
- |
3 |
672 |
|
View Full Product Details