Gene Transcripts
- Gene Symbol
- Sh3kbp1
- Entrez Gene ID
- 58194
- Gene Name
- SH3-domain kinase binding protein 1
- Gene Aliases
- 1200007H22Rik, 1700125L08Rik, 5830464D22Rik, AI447724, IN85, Ruk, Seta
- Chromosome Location
- Chr. X: 159627029 - 159975920 on Build GRCm38
- UniGene ID
- Mm.286495
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135727.2 |
NP_001129199.1
|
17, 18 |
2375 |
|
NM_001135728.2 |
NP_001129200.1
|
12, 13 |
1377 |
|
NM_001290661.1 |
NP_001277590.1
|
6, 7 |
1057 |
|
NM_001290664.1 |
NP_001277593.1
|
5, 6 |
928 |
|
NM_021389.6 |
NP_067364.2
|
16, 17 |
1940 |
|
GenBank mRNA |
AK004636.1 |
- |
16, 17 |
1940 |
|
AK007283.1 |
- |
6, 7 |
1043 |
|
AK028299.1 |
- |
12, 13 |
1381 |
|
AK030839.1 |
- |
16, 17 |
1944 |
|
AK049182.1 |
- |
17, 18 |
2376 |
|
AK052260.1 |
- |
5, 6 |
603 |
|
AK080930.1 |
- |
17, 18 |
2241 |
|
AK142295.1 |
- |
6, 7 |
831 |
|
AK161512.1 |
- |
6, 7 |
1058 |
|
AK161735.1 |
- |
12, 13 |
1458 |
|
AK172022.1 |
- |
17, 18 |
2356 |
|
AK202551.1 |
- |
|
12 |
|
BC034646.1 |
- |
16, 17 |
1945 |
|
BC052863.1 |
- |
16, 17 |
1885 |
|
View Full Product Details