Gene Transcripts

Gene Symbol
Hgh1
Entrez Gene ID
59053
Gene Name
HGH1 homolog
Gene Aliases
Brp16, D15Ertd741e, Fam203a, MNCb-5873
Chromosome Location
Chr. 15: 76368860 - 76371411 on Build GRCm38
UniGene ID
Mm.133202
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021555.2 NP_067530.2 4 946
GenBank mRNA AB041552.1 - 4 946
AK085815.1 - 4 940
AK178524.1 - 2 225
BC061251.1 - 4 910 30249656

View Full Product Details