Gene Transcripts

Gene Symbol
Tnmd
Entrez Gene ID
64103
Gene Name
tenomodulin
Gene Aliases
1110017I01Rik, Bricd4, ChM1L, TeM
Chromosome Location
Chr. X: 133851008 - 133865577 on Build GRCm38
UniGene ID
Mm.46221
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022322.2 NP_071717.1 3 456
GenBank mRNA AB055422.1 - 3 263
AF191768.1 - 3 402
AF219993.1 - 3 329
AF291655.1 - 3 481
AK003748.1 - 3 342
AK014761.1 - 3 295
BC006919.1 - 3 265 3601223
BC049944.1 - 3 456 2811628

View Full Product Details