Gene Transcripts
- Gene Symbol
- Ilf2
- Entrez Gene ID
- 67781
- Gene Name
- interleukin enhancer binding factor 2
- Gene Aliases
- 6230405A16Rik, TEG-267, Tex261, Tex267
- Chromosome Location
- Chr. 3: 90476201 - 90488379 on Build GRCm38
- UniGene ID
- Mm.227258
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026374.3 |
NP_080650.1
|
9, 10 |
687 |
|
GenBank mRNA |
AK012364.1 |
- |
9, 10 |
674 |
|
AK013858.1 |
- |
9, 10 |
733 |
|
AK078003.1 |
- |
9, 10 |
674 |
|
AK135449.1 |
- |
9, 10 |
762 |
|
AK135539.1 |
- |
9, 10 |
674 |
|
AK157129.1 |
- |
9, 10 |
742 |
|
AK212445.1 |
- |
1, 2 |
23 |
|
BC004033.1 |
- |
1, 2 |
54 |
|
BC024718.1 |
- |
9, 10 |
687 |
5012365 |
View Full Product Details