Gene Transcripts

Gene Symbol
Spata4
Entrez Gene ID
69281
Gene Name
spermatogenesis associated 4
Gene Aliases
1700001N01Rik, Srg2, TSARG2
Chromosome Location
Chr. 8: 54600781 - 54610098 on Build GRCm38
UniGene ID
Mm.45833
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133711.3 NP_598472.2 5 770
GenBank mRNA AF395083.2 - 5 770
AF530515.1 - 5 770
AK005612.1 - 5 770
BC048650.1 - 5 770 6772684

View Full Product Details