Gene Transcripts
- Gene Symbol
- Tecpr1
- Entrez Gene ID
- 70381
- Gene Name
- tectonin beta-propeller repeat containing 1
- Gene Aliases
- 2210010N04Rik, mKIAA1358
- Chromosome Location
- Chr. 5: 144194442 - 144223608 on Build GRCm38
- UniGene ID
- Mm.239412
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_027410.1 |
NP_081686.1
|
23, 24 |
3380 |
|
GenBank mRNA |
AB093295.1 |
- |
22, 23 |
3549 |
|
AK008701.1 |
- |
6, 7 |
618 |
|
AK032858.1 |
- |
23, 24 |
3354 |
|
AK169888.1 |
- |
23, 24 |
3327 |
|
AK170108.1 |
- |
23, 24 |
3327 |
|
AK199688.1 |
- |
1, 2 |
75 |
|
AK202952.1 |
- |
3, 4 |
321 |
|
AK211137.1 |
- |
1, 2 |
75 |
|
AK211310.1 |
- |
3, 4 |
347 |
|
BC004058.1 |
- |
6, 7 |
666 |
|
BC025567.1 |
- |
10, 11 |
1125 |
|
BC046968.1 |
- |
23, 24 |
3380 |
6306206 |
View Full Product Details