Gene Transcripts

Gene Symbol
Rufy2
Entrez Gene ID
70432
Gene Name
RUN and FYVE domain-containing 2
Gene Aliases
2610111M19Rik, AI844453, Denn, LZ-FYVE, ZFYVE13
Chromosome Location
Chr. 10: 62980223 - 63018742 on Build GRCm38
UniGene ID
Mm.234587
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027425.3 NP_081701.2 4 470
GenBank mRNA AK011847.1 - 4 440
AK083205.1 - 4 455
AK173195.1 - 4 431
BC017645.1 - 4 432
BC038265.1 - 4 411
BC137736.1 - 4 419 8860755
BC137737.1 - 4 419 8860756

View Full Product Details