Gene Transcripts

Gene Symbol
Ankib1
Entrez Gene ID
70797
Gene Name
ankyrin repeat and IBR domain containing 1
Gene Aliases
2310061P20Rik, 4631416I11Rik, AW494740, C80642, mKIAA1386
Chromosome Location
Chr. 5: 3689999 - 3803124 on Build GRCm38
UniGene ID
Mm.130148
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003909.4 NP_001003909.2 9 1903
NM_001289527.1 NP_001276456.1 9 1903
NM_001289528.1 NP_001276457.1 8 1785
NM_001289529.1 NP_001276458.1 8 1375
GenBank mRNA AK129343.1 - 9 1704
BC020475.1 - 9 1437
BC089031.1 - 8 1354
BC145510.1 - 8 1536
BC145511.1 - 9 1654 9054074
BC150792.1 - 9 1654 9087703

View Full Product Details