Gene Transcripts

Gene Symbol
Fsip1
Entrez Gene ID
71313
Gene Name
fibrous sheath-interacting protein 1
Gene Aliases
1700012M13Rik, 4933432K11Rik
Chromosome Location
Chr. 2: 118130261 - 118256987 on Build GRCm38
UniGene ID
Mm.85046
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027759.3 NP_082035.1 7, 8 982
GenBank mRNA AF448787.1 - 7, 8 982
AK017026.1 - 7, 8 971
BC125024.1 - 7, 8 874 40056533

View Full Product Details