Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001168371.2 | NP_001161843.1 | 12 | 1976 | |
NM_001168372.2 | NP_001161844.1 | 12 | 1973 | ||
NM_001168373.2 | NP_001161845.1 | 12 | 1970 | ||
NM_027804.4 | NP_082080.3 | 12 | 1973 | ||
NM_145407.3 | NP_663382.2 | 11 | 1901 | ||
GenBank mRNA | AK043088.1 | - | 11 | 1737 | |
AK122396.2 | - | 11 | 1759 | ||
AK144099.1 | - | 7 | 1005 | ||
AK146504.1 | - | 12 | 1799 | ||
AK148412.1 | - | 7 | 962 | ||
AK159473.1 | - | 12 | 1795 | ||
AK160807.1 | - | 12 | 1799 | ||
AK171942.1 | - | 12 | 1802 | ||
BC006916.1 | - | 4 | 508 | ||
BC046824.1 | - | 3 | 436 | ||
BC060613.1 | - | 12 | 1901 | 6400986 |