Gene Transcripts
- Gene Symbol
- Fam13c
- Entrez Gene ID
- 71721
- Gene Name
- family with sequence similarity 13, member C
- Gene Aliases
- 1200015N20Rik, C030038O19Rik, mKIAA1796
- Chromosome Location
- Chr. 10: 70440668 - 70558732 on Build GRCm38
- UniGene ID
- Mm.477813
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK004797.1 |
- |
14 |
1877 |
|
View Full Product Details