Gene Transcripts

Gene Symbol
Golim4
Entrez Gene ID
73124
Gene Name
golgi integral membrane protein 4
Gene Aliases
3110027H23Rik, GIMPC, GPP130, Golph4, P138
Chromosome Location
Chr. 3: 75876183 - 75956949 on Build GRCm38
UniGene ID
Mm.249232
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291069.1 NP_001277998.1 10 1948
NM_175193.6 NP_780402.1 9 1864
GenBank mRNA AK033730.1 - 9 1851
AK048434.1 - 10 1775
AK078928.1 - 10 1948
BC086642.1 - 9 1696 6836803
BC089354.1 - 9 1746 6837211

View Full Product Details