Gene Transcripts

Gene Symbol
Hvcn1
Entrez Gene ID
74096
Gene Name
hydrogen voltage-gated channel 1
Gene Aliases
0610039P13Rik, AI450555, BTS, HV1, Vsop., mVSOP
Chromosome Location
Chr. 5: 122206807 - 122242297 on Build GRCm38
UniGene ID
Mm.28804
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042489.2 NP_001035954.1 6 937
NM_028752.3 NP_083028.1 5 799
GenBank mRNA AK002854.1 - 5 800
AK154880.1 - 6 829
AK155123.1 - 6 774
AK170422.1 - 6 798
AK191370.1 - 1 111
BC021548.1 - 6 768 4913027
BY703352.1 - 5 793

Gene Symbol
Tctn1
Entrez Gene ID
654470
Gene Name
tectonic family member 1
Gene Aliases
G730031O11Rik, Tect1
Chromosome Location
Chr. 5: 122239495 - 122264489 on Build GRCm38
UniGene ID
Mm.28804
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK135183.1 - 1597

View Full Product Details