Gene Transcripts
- Gene Symbol
- Fastkd2
- Entrez Gene ID
- 75619
- Gene Name
- FAST kinase domains 2
- Gene Aliases
- 2810421I24Rik
- Chromosome Location
- Chr. 1: 63730599 - 63753385 on Build GRCm38
- UniGene ID
- Mm.30965
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172422.3 |
NP_766010.1
|
8, 9 |
1712 |
|
GenBank mRNA |
AK035322.1 |
- |
8, 9 |
1712 |
|
AK084971.1 |
- |
8, 9 |
1687 |
|
AK129251.1 |
- |
6, 7 |
2889 |
|
AK148224.1 |
- |
|
1696 |
|
AK153916.1 |
- |
7, 8 |
1655 |
|
AK166378.1 |
- |
8, 9 |
1697 |
|
AK167714.1 |
- |
7, 8 |
1968 |
|
BC008271.1 |
- |
7, 8 |
1653 |
3603187 |
BC057208.1 |
- |
7, 8 |
1646 |
5328766 |
BC080813.1 |
- |
7, 8 |
1652 |
6394260 |
View Full Product Details