Gene Transcripts
- Gene Symbol
- Abcb5
- Entrez Gene ID
- 77706
- Gene Name
- ATP-binding cassette, sub-family B (MDR/TAP), member 5
- Gene Aliases
- 9230106F14Rik
- Chromosome Location
- Chr. 12: 118867486 - 118966421 on Build GRCm38
- UniGene ID
- Mm.261496
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_029961.2 |
NP_084237.1
|
13 |
1520 |
|
GenBank mRNA |
AY766239.1 |
- |
13 |
1484 |
|
View Full Product Details