Gene Transcripts
- Gene Symbol
- Fam53b
- Entrez Gene ID
- 77938
- Gene Name
- family with sequence similarity 53, member B
- Gene Aliases
- A930008G19Rik, mKIAA0140
- Chromosome Location
- Chr. 7: 132712078 - 132813196 on Build GRCm38
- UniGene ID
- Mm.225649
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_175268.4 |
NP_780477.2
|
2, 3 |
306 |
|
NM_212473.1 |
NP_997638.1
|
2, 3 |
369 |
|
GenBank mRNA |
AK020830.1 |
- |
2, 3 |
307 |
|
AK051299.1 |
- |
2, 3 |
370 |
|
AK147350.1 |
- |
2, 3 |
702 |
|
AK147725.1 |
- |
2, 3 |
498 |
|
AK156629.1 |
- |
2, 3 |
269 |
|
AK165022.1 |
- |
|
363 |
|
AK180944.1 |
- |
1, 2 |
123 |
|
AK186433.1 |
- |
1, 2 |
123 |
|
AK187644.1 |
- |
1, 2 |
123 |
|
AK195672.1 |
- |
1, 2 |
74 |
|
AK207193.1 |
- |
1, 2 |
123 |
|
BC035523.1 |
- |
2, 3 |
301 |
5400742 |
View Full Product Details