Gene Transcripts
- Gene Symbol
- Rbm5
- Entrez Gene ID
- 83486
- Gene Name
- RNA binding motif protein 5
- Gene Aliases
- D030069N10Rik
- Chromosome Location
- Chr. 9: 107740495 - 107771008 on Build GRCm38
- UniGene ID
- Mm.259197
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_148930.3 |
NP_683732.1
|
9, 10 |
851 |
|
GenBank mRNA |
AJ309168.1 |
- |
8, 9 |
686 |
|
AK134183.1 |
- |
9, 10 |
846 |
|
AK146880.1 |
- |
9, 10 |
846 |
|
BC005719.1 |
- |
4, 5 |
456 |
|
BC019766.1 |
- |
9, 10 |
796 |
|
BC023854.1 |
- |
9, 10 |
836 |
5346780 |
BC031899.1 |
- |
9, 10 |
836 |
5362121 |
BC042419.1 |
- |
7, 8 |
4087 |
|
BC043058.1 |
- |
9, 10 |
788 |
6419695 |
BC054729.1 |
- |
9, 10 |
839 |
6419758 |
BC054820.1 |
- |
7, 8 |
3565 |
|
BC060985.1 |
- |
8, 9 |
755 |
|
View Full Product Details