Gene Transcripts

Gene Symbol
Chn1
Entrez Gene ID
108699
Gene Name
chimerin 1
Gene Aliases
0610007I19Rik, 0710001E19Rik, 1700112L09Rik, 2900046J01Rik, AI413815, ARHGAP2
Chromosome Location
Chr. 2: 73610660 - 73775367 on Build GRCm38
UniGene ID
Mm.257073
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113246.2 NP_001106717.2 13 1649
NM_001166603.1 NP_001160075.1 12 1475
NM_001166604.1 NP_001160076.1 6 1287
NM_029716.3 NP_083992.1 6 850
NM_175752.3 NP_786928.2 7 1461
GenBank mRNA AB264771.1 - 6 1111
AB264772.1 - 12 1172
AB264773.1 - 3818
AB264774.1 - 5 1251
AF332069.1 - 7 969
AF332070.1 - 7 969
AK007182.1 - 6 850
AK032630.1 - 7 1419
AK049943.1 - 7 1405
AK075606.1 - 7 1423
AK147686.1 - 7 1405
BC010825.1 - 7 1436 4219273
BC024796.1 - 7 1412 5360438
BC025023.1 - 7 1392 5364554
BC054770.1 - 13 1401 6415204

View Full Product Details